missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydro-2-(1H)-quinolinone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3,4-Dihydro-2-(1H)-quinolinone may be employed as medium supplement in the culture medium of Pseudomonas ayucida during enrichment culture experiments. It is used to prepare potent bicyclic peptide deformylase inhibitors with antibacterial effects. It is also used to synthesize substituted iminopiperidines as inhibitors of human nitric oxide synthase isoforms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dihydro-2-(1H)-quinolinone may be employed as medium supplement in the culture medium of Pseudomonas ayucida during enrichment culture experiments. It is used to prepare potent bicyclic peptide deformylase inhibitors with antibacterial effects. It is also used to synthesize substituted iminopiperidines as inhibitors of human nitric oxide synthase isoforms.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
3,4-Dihydro-2-(1H)-quinolinone may be employed as medium supplement in the culture medium of Pseudomonas ayucida during enrichment culture experiments. It is used to prepare potent bicyclic peptide deformylase inhibitors with antibacterial effects. It is also used to synthesize substituted iminopiperidines as inhibitors of human nitric oxide synthase isoforms.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
Melting Point | 165°C to 167°C |
Quantity | 25 g |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble in water. |
IUPAC Name | 1,2,3,4-tetrahydroquinolin-2-one |
Formula Weight | 147.18 |
Percent Purity | 98% |
Chemical Name or Material | 3,4-Dihydro-2-(1H)-quinolinone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'