Läs mer
3,4-Dihydro-6,7-dimethoxy-4-methyl-3-oxo-2-quinoxalinepropanoic Acid, TRC
Produktkod. 30369922
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30369922 5 mg
30376733 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30369922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4490655MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Dihydro-6,7-dimethoxy-4-methyl-3-oxo-2-quinoxalinepropanoic Acid
CAS 132788-56-8
Molekylformel C14 H16 N2 O5
Kvantitet 5 mg
Synonym 6,7-Dimethoxy-4-methyl-3-oxo-3,4-dihydroquinoxaline-2-propionic Acid
InChI formel InChI=1S/C14H16N2O5/c1-16-10-7-12(21-3)11(20-2)6-9(10)15-8(14(16)19)4-5-13(17)18/h6-7H,4-5H2,1-3H3,(H,17,18)
LEDER COc1cc2N=C(CCC(=O)O)C(=O)N(C)c2cc1OC
IUPAC-namn 3-(6,7-dimethoxy-4-methyl-3-oxoquinoxalin-2-yl)propanoic acid
Molekylvikt (g/mol) 292.29
Formel vikt 292.1059