Läs mer
3,4-Dihydro Ivermectin (Mixture of Diastereomers) (>90%), TRC
Produktkod. 30009935
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009935 10 mg
30005464 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009935 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44682010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74567-01-4
Molekylformel C48 H76 O14
Molekylvikt (g/mol) 877.11
Synonym Avermectin A1a, 5-O-demethyl-3,4,22,23-tetrahydro-
LEDER CC[C@H](C)[C@H]1O[C@]2(CC[C@@H]1C)C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/CO[C@@H]7[C@H](O)C(C)C[C@@H](C(=O)O3)[C@]67O)O2
Specifikationer
Kemiskt namn eller material 3,4-Dihydro Ivermectin (mixture of diastereomers)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C48H76O14/c1-11-25(2)43-28(5)17-18-47(62-43)23-34-20-33(61-47)16-15-27(4)42(26(3)13-12-14-32-24-55-45-40(49)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-10)44(31(8)57-39)60-38-21-36(53-9)41(50)30(7)56-38/h12-15,25-26,28-31,33-45,49-50,52H,11,16-24H2,1-10H3/b13-12+,27-15+,32-14+/t25-,26-,28-,29?,30-,31-,33+,34-,35-,36-,37-,38-,39-,40+,41-,42-,43+,44-,45+,47+,48+/m0/s1
Formel vikt 876.5235
Procent renhet >95%