Läs mer
3,4-Dihydro Naratriptan Oxalate, TRC
Produktkod. 30369046
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30369046 100 mg
1 options
Produktkod. 30369046 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD449202100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Dihydro Naratriptan Oxalate
CAS 121679-21-8
Molekylformel C17H23N3O2S . C2H2O4
Kvantitet 100 mg
Synonym (Naratriptan Impurity B), N-Methyl-3-(1,2,3,6-tetrahydro-1-methyl-4-pyridinyl)-1H-indole-5-ethanesulfonamide Ethanedioate
InChI formel InChI=1S/C17H23N3O2S.C2H2O4/c1-18-23(21,22)10-7-13-3-4-17-15(11-13)16(12-19-17)14-5-8-20(2)9-6-14;3-1(4)2(5)6/h3-5,11-12,18-19H,6-10H2,1-2H3;(H,3,4)(H,5,6)
LEDER CNS(=O)(=O)CCc1ccc2[nH]cc(C3=CCN(C)CC3)c2c1.OC(=O)C(=O)O
Molekylvikt (g/mol) 423.5
Formel vikt 423.1464