Läs mer
3,4-Dihydroxy-5-nitrobenzoic Acid, TRC
Produktkod. 30386360
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30386360 250 mg -
30381283 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30386360 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD453235250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84211-30-3
Molekylformel C7H5NO6
Molekylvikt (g/mol) 199.12
Synonym 3-Nitro-4,5-dihydroxybenzoic Acid
LEDER Cl.CNC(C)Cc1cc(OC)c(O)cc1
Specifikationer
Kemiskt namn eller material 3,4-Dihydroxy-5-nitrobenzoic Acid
Kvantitet 250 mg
InChI formel InChI=1S/C7H5NO6/c9-5-2-3(7(11)12)1-4(6(5)10)8(13)14/h1-2,9-10H,(H,11,12)