Läs mer
3,4-Dimethoxy-2-(hydroxymethyl)pyridine, Mikromol™
Produktkod. 30052345
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052345 25 mg
17911436 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052345 Leverantör Mikromol Leverantörsnummer MM0099.140025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3,4-Dimethoxy-2-(hydroxymethyl)pyridine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Dimethoxy-2-(hydroxymethyl)pyridine
CAS 72830-08-1
Analyt- eller komponentnamn 3,4-Dimethoxy-2-(hydroxymethyl)pyridine
Frakt skick Room Temperature
Molekylformel C8 H11 N O3
Certifikat och efterlevnad ISO 17025
Synonym 2-Pyridinemethanol, 3,4-dimethoxy-, 2-Hydroxymethyl-3,4-dimethoxypyridine, 3,4-Dimethoxy-2-(hydroxymethyl)pyridine, 3,4-Dimethoxy-2-pyridinemethanol
InChI formel InChI=1S/C8H11NO3/c1-11-7-3-4-9-6(5-10)8(7)12-2/h3-4,10H,5H2,1-2H3
LEDER COc1ccnc(CO)c1OC
IUPAC-namn (3,4-dimethoxypyridin-2-yl)methanol
Molekylvikt (g/mol) 169.18
Kvantitet 25 mg
Formel vikt 169.0739 g/mol