missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzamidoxime, 97%, Thermo Scientific™

Catalog Number p-7034517
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1053058-99-3
C9H12N2O3
196.21
MFCD06147747,MFCD09842202
REKFOHFJHBBXOR-UHFFFAOYSA-N
3,4-dimethoxybenzamidoxime, z-n'-hydroxy-3,4-dimethoxybenzene-1-carboximidamide, 3,4-dimethoxyphenyl hydroxyimino methylamine, n'-hydroxy-3,4-dimethoxybenzimidamide, benzenecarboximidamide, n-hydroxy-3,4-dimethoxy, n'-hydroxy-3,4-dimethoxybenzene-1-carboximidamide, n'∼1∼-hydroxy-3,4-dimethoxy-1-benzenecarboximidamide, e-n'-hydroxy-3,4-dimethoxybenzenecarboximidamide
9723450
N'-hydroxy-3,4-dimethoxybenzenecarboximidamide
COC1=C(OC)C=C(C=C1)C(N)=NO
This item is not returnable. View return policy
1053058-99-3
C9H12N2O3
196.21
MFCD06147747,MFCD09842202
REKFOHFJHBBXOR-UHFFFAOYSA-N
3,4-dimethoxybenzamidoxime, z-n'-hydroxy-3,4-dimethoxybenzene-1-carboximidamide, 3,4-dimethoxyphenyl hydroxyimino methylamine, n'-hydroxy-3,4-dimethoxybenzimidamide, benzenecarboximidamide, n-hydroxy-3,4-dimethoxy, n'-hydroxy-3,4-dimethoxybenzene-1-carboximidamide, n'∼1∼-hydroxy-3,4-dimethoxy-1-benzenecarboximidamide, e-n'-hydroxy-3,4-dimethoxybenzenecarboximidamide
9723450
N'-hydroxy-3,4-dimethoxybenzenecarboximidamide
COC1=C(OC)C=C(C=C1)C(N)=NO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 196.21
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxybenzamidoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'