Läs mer
3,4-Dimethoxyphenyl isothiocyanate, 99%, Thermo Scientific™
Produktkod. 11439387
Change view
Kvantitet:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11439387 25 g 25g
11419387 1 g 1g
11429387 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11439387 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B21415.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 33904-04-0
Molekylformel C9H9NO2S
Molekylvikt (g/mol) 195.236
MDL-nummer MFCD00041075
InChI-nyckel LHPZZVZPOZPDDB-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenyl isothiocyanate, 3,4-dimethoxyphenylisothiocyanate, 4-isothiocyanato-1,2-dimethoxy-benzene, isothiocyanic acid 3,4-dimethoxyphenyl ester, 3,4-dimethoxybenzenisothiocyanate, acmc-1aeuy, 3,4 dimethoxyphenylisothiocyanate, lhpzzvzpozpddb-uhfffaoysa, #, 3,4-dimethoxy phenyl isothiocyanate
PubChem CID 141856
IUPAC-namn 4-isothiocyanato-1,2-dimethoxybenzene
LEDER COC1=C(C=C(C=C1)N=C=S)OC
Specifikationer
Kemiskt namn eller material 3,4-Dimethoxyphenyl isothiocyanate
Smältpunkt 47°C to 49°C
Kokpunkt 171°C to 172°C (7 mmHg)
Kvantitet 25 g
UN-nummer UN2923
Beilstein 2726948
Känslighet Moisture sensitive
Formel vikt 195.24
Procent renhet 99%