Läs mer
3,4-Dimethyl-1-(1-chloroethyl)benzene (>85%), TRC
Produktkod. 30179950
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179950 50 mg
30180362 250 mg
30180424 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47183350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 104245-83-2
Molekylformel C10 H13 Cl
Molekylvikt (g/mol) 168.66
Synonym Medetomidine Impurity 30
IUPAC-namn 4-(1-chloroethyl)-1,2-dimethylbenzene
LEDER ClC(C)C1=CC=C(C)C(C)=C1
Specifikationer
Kemiskt namn eller material 3,4-Dimethyl-1-(1-chloroethyl)benzene
Kvantitet 50 mg
InChI formel InChI=1S/C10H13Cl/c1-7-4-5-10(9(3)11)6-8(7)2/h4-6,9H,1-3H3
Formel vikt 168.07