Läs mer
3,4-Dimethyl-1,2-cyclopentanedione, TRC
Produktkod. 30356910
Change view
Kvantitet:
1 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356910 2.5 g
30350887 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30356910 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2663952.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13494-06-9
Molekylformel C7H10O2
Molekylvikt (g/mol) 126.15
IUPAC-namn 3,4-dimethylcyclopentane-1,2-dione
LEDER CC1CC(=O)C(=O)C1C
Specifikationer
Kemiskt namn eller material 3,4-Dimethyl-1,2-cyclopentanedione
Kvantitet 2.5 g
InChI formel InChI=1S/C7H10O2/c1-4-3-6(8)7(9)5(4)2/h4-5H,3H2,1-2H3
Formel vikt 126.0681