Läs mer
3,4-Dimethyl-3-cyclopentene-1-carboxylic Acid, TRC
Produktkod. 30380472
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30380472 2.5 mg 2.5mg
30381216 25 mg -
30372516 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30380472 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD6152452.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 768-38-7
Molekylformel C8H12O2
Molekylvikt (g/mol) 140.18
Synonym Rosiglitazone Related Compound 1
IUPAC-namn 3,4-dimethylcyclopent-3-ene-1-carboxylic acid
LEDER CC1=C(C)CC(C1)C(=O)O
Specifikationer
Kemiskt namn eller material 3,4-Dimethyl-3-cyclopentene-1-carboxylic Acid
Kvantitet 2.5 mg
InChI formel InChI=1S/C8H12O2/c1-5-3-7(8(9)10)4-6(5)2/h7H,3-4H2,1-2H3,(H,9,10)
Formel vikt 140.0837