missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7024328
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
528-45-0
C7H4N2O6
212.12
MFCD00007255
OMVRRHJJQILIJX-UHFFFAOYSA-N
3,4-dinitrobenzoic acid, benzoic acid, 3,4-dinitro, unii-0l3n4t0gpd, ccris 3128, 3,4-dinitrobenzioc acid, 0l3n4t0gpd, 3,4-dinitrobenzoate, 3,4-di-nitro-benzoic acid, zlchem 522, pubchem4956
10709
3,4-dinitrobenzoic acid
OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O
This item is not returnable. View return policy
528-45-0
C7H4N2O6
212.12
MFCD00007255
OMVRRHJJQILIJX-UHFFFAOYSA-N
3,4-dinitrobenzoic acid, benzoic acid, 3,4-dinitro, unii-0l3n4t0gpd, ccris 3128, 3,4-dinitrobenzioc acid, 0l3n4t0gpd, 3,4-dinitrobenzoate, 3,4-di-nitro-benzoic acid, zlchem 522, pubchem4956
10709
3,4-dinitrobenzoic acid
OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 163°C to 167°C
Quantity 5 g
Beilstein 2055333
Merck Index 14,3275
Formula Weight 212.12
Percent Purity 99%
Chemical Name or Material 3,4-Dinitrobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'