Läs mer
3,4-Epoxycyclohexylmethyl 3,4-Epoxycyclohexaneca, TRC
Produktkod. 30377976
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30377976 1 g
1 options
Produktkod. 30377976 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5859201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Epoxycyclohexylmethyl 3,4-Epoxycyclohexaneca
CAS 2386-87-0
Molekylformel C14 H20 O4
Kvantitet 1 g
Synonym (3,4-Epoxycyclohexane)methyl 3,4-epoxycyclohexylcarboxylate, (3,4-Epoxycyclohexane)methyl 3',4'-epoxycyclohexyl-carboxylate, (3,4-Epoxycyclohexyl)methyl 3,4-epoxycyclohexylcarboxylate, (3',4'-Epoxycyclohexane)methyl 3,4-epoxycyclohexanecarboxylate, (3',4'-Epoxycyclohexylmethyl) 3,4-epoxycyclohexanecarboxylate, 3,4-Epoxycyclohexanecarboxylic acid (3,4-epoxycyclohexylmethyl) ester, 3,4-Epoxycyclohexanecarboxylic acid (3',4'-epoxycyclohexyl)methyl ester, 3,4-Epoxycyclohexanemethyl 3,4-epoxycyclohexanecarboxylate, 3,4-Epoxycyclohexenylmethyl 3',4'-epoxycyclohexenecarboxylate, 3,4-Epoxycyclohexylmethyl 3,4-epoxycyclohexanecarboxylate, 3,4-Epoxycyclohexylmethyl 3',4'-epoxycyclohexanecarboxylate, 3,4-Epoxycyclohexylmethyl 3',4'-epoxycyclohexylcarboxylate, 3,4-Epoxycyclohexylmethylene-(3,4-epoxycyclohexane) carboxylate, 7-Oxabicyclo[4.1.0]hept-3-yl-methyl 7-oxabicyclo[4.1.0]heptane-3-carboxylate, 7-Oxabicyclo[4.1.0]heptan-4-ylmethyl 7-oxabicyclo[4.1.0]heptane-4-carboxylate, Chissonox 221 monomer, EPOX, UP 632 monomer, UVA 1500 monomer, 7-Oxabicyclo[4.1.0]heptane-3-carboxylic acid, 7-oxabicyclo[4.1.0]hept-3-ylmethyl ester
InChI formel InChI=1S/C14H20O4/c15-14(9-2-4-11-13(6-9)18-11)16-7-8-1-3-10-12(5-8)17-10/h8-13H,1-7H2
LEDER O=C(OCC1CCC2OC2C1)C3CCC4OC4C3
IUPAC-namn 7-oxabicyclo[4.1.0]heptan-4-ylmethyl 7-oxabicyclo[4.1.0]heptane-4-carboxylate
Molekylvikt (g/mol) 252.31
Formel vikt 252.1362