Läs mer
3-(4-Ethylphenyl)-2,2-dimethylpropanal, TRC
Produktkod. 30368792
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30368792 25 g
1 options
Produktkod. 30368792 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE67964025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(4-Ethylphenyl)-2,2-dimethylpropanal
CAS 67634-15-5
Molekylformel C13H18O
Kvantitet 25 g
Synonym Floralozone, Florazon, Seawind Aldehyde E, alpha,alpha-Dimethyl-p-ethylphenylpropanal
InChI formel InChI=1S/C13H18O/c1-4-11-5-7-12(8-6-11)9-13(2,3)10-14/h5-8,10H,4,9H2,1-3H3
LEDER CCc1ccc(CC(C)(C)C=O)cc1
Molekylvikt (g/mol) 190.28
Formel vikt 190.1358