Läs mer
3-(4-Formylaminobutyryl)pyridine, TRC
Produktkod. 30006860
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006860 25 mg
30006297 5 mg
30007264 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006860 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF69550025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 887355-56-8
Molekylformel C10H12N2O2
Molekylvikt (g/mol) 192.21
Synonym Nicotine Impurity 1
LEDER O=CNCCCC(=O)c1cccnc1
Specifikationer
Kemiskt namn eller material 3-(4-Formylaminobutyryl)pyridine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C10H12N2O2/c13-8-12-6-2-4-10(14)9-3-1-5-11-7-9/h1,3,5,7-8H,2,4,6H2,(H,12,13)
Formel vikt 192.0899
Procent renhet >95%