Läs mer
3-(4-Methylaminobutyryl)pyridine-1,2',3',4',5',6'-13C6, Dihydrochloride, TRC
Produktkod. 30136972
Change view
Kvantitet:
0.25 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136972 0.25 mg
30005569 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136972 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2857520.25MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246819-72-6
Molekylformel C413C6H16Cl2N2O
Molekylvikt (g/mol) 257.11
Synonym 4-Methylamino-1-(3-pyridyl)-1-butanone-13C6 DiHydrochloride, Pseudooxynicotine 13C6 DiHydrochloride
LEDER O=[13C](CCCNC)[13C]1=[13C]([H])[13C]([H])=[13C]([H])N=[13C]1[H]
Specifikationer
Kemiskt namn eller material 3-(4-Methylaminobutyryl)pyridine-1,2',3',4',5',6'-13C6, Dihydrochloride
Kvantitet 0.25 mg
InChI formel InChI=1S/C10H14N2O/c1-11-6-3-5-10(13)9-4-2-7-12-8-9/h2,4,7-8,11H,3,5-6H2,1H3/i2+1,4+1,7+1,8+1,9+1,10+1