missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methylpiperazin-1-yl)benzoic acid, ≥97%, Thermo Scientific™

Catalog Number p-3175680
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
215309-01-6
C12H16N2O2
220.27
MFCD06742238
HALATUFUWLWCQV-UHFFFAOYSA-N
3-4-methylpiperazin-1-yl benzoic acid, 3-4-methylpiperazin-1-yl benzoicacid, benzoicacid, 3-4-methyl-1-piperazinyl, zlchem 853, pubchem12997, acmc-209fli, r-1-fmoc-3-hydroxypyrrolidine, 3-4-methylpiperazinyl benzoic acid, 3-4-methylpiperazinyl-benzoic acid, 3-4-methyl-1-piperazinyl benzoic acid
4741681
3-(4-methylpiperazin-1-yl)benzoic acid
CN1CCN(CC1)C1=CC=CC(=C1)C(O)=O
This item is not returnable. View return policy
215309-01-6
C12H16N2O2
220.27
MFCD06742238
HALATUFUWLWCQV-UHFFFAOYSA-N
3-4-methylpiperazin-1-yl benzoic acid, 3-4-methylpiperazin-1-yl benzoicacid, benzoicacid, 3-4-methyl-1-piperazinyl, zlchem 853, pubchem12997, acmc-209fli, r-1-fmoc-3-hydroxypyrrolidine, 3-4-methylpiperazinyl benzoic acid, 3-4-methylpiperazinyl-benzoic acid, 3-4-methyl-1-piperazinyl benzoic acid
4741681
3-(4-methylpiperazin-1-yl)benzoic acid
CN1CCN(CC1)C1=CC=CC(=C1)C(O)=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 3-(4-Methylpiperazin-1-yl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'