Läs mer
3-(4-Piperidinyl)-1,2-benzisoxazole N-Carbamic Acid Phenyl Ester, TRC
Produktkod. 30427653
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30427653 10 mg
1 options
Produktkod. 30427653 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP48200510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(4-Piperidinyl)-1,2-benzisoxazole N-Carbamic Acid Phenyl Ester
CAS 84163-21-3
Molekylformel C19H18N2O3
Kvantitet 10 mg
Synonym 4-(1,2-Benzisoxazol-3-yl)-1-piperidinecarboxylic Acid Phenyl Ester
InChI formel InChI=1S/C19H18N2O3/c22-19(23-15-6-2-1-3-7-15)21-12-10-14(11-13-21)18-16-8-4-5-9-17(16)24-20-18/h1-9,14H,10-13H2
LEDER O=C(Oc1ccccc1)N2CCC(CC2)c3noc4ccccc34
IUPAC-namn phenyl 4-(1,2-benzoxazol-3-yl)piperidine-1-carboxylate
Molekylvikt (g/mol) 322.36
Formel vikt 322.1317