Läs mer
3-(4-Piperidinyloxy)benzonitrile hydrochloride, 97%, Thermo Scientific™
Produktkod. 15439195
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15439195 250 mg 250mg
15429195 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15439195 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H51143.MD
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 950649-07-7
Molekylformel C12H15ClN2O
Molekylvikt (g/mol) 238.715
MDL-nummer MFCD11977714
InChI-nyckel YRNQJNMHJHPFRA-UHFFFAOYSA-N
Synonym 3-4-piperidinyloxy benzonitrile hydrochloride, 3-4-piperidinyloxy benzonitrile hcl, 3-4-piperidinyloxy-benzonitrile hcl, 4-3-cyanophenyloxy piperidine hydrochloride, 3-piperidin-4-yloxy-benzonitrile hydrochloride, 3-piperidin-4-yloxy benzonitrile hydrochloride, 3-piperidin-4-yl oxy benzonitrile-hydrogen chloride 1/1
PubChem CID 53433745
IUPAC-namn 3-piperidin-4-yloxybenzonitrile;hydrochloride
LEDER C1CNCCC1OC2=CC=CC(=C2)C#N.Cl
Specifikationer
Kemiskt namn eller material 3-(4-Piperidinyloxy)benzonitrile hydrochloride
Kvantitet 250 mg
Formel vikt 238.7
Procent renhet 97%