missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Pyridyl)propanoic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 229.0°C to 233.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Quantity | 5 g |
Formula Weight | 151.17 |
Percent Purity | 95% |
Chemical Name or Material | 3-(4-Pyridyl)propanoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'