Läs mer
3,4-Secocholest-5-ene-3,4-dioic Acid Dimethyl Ester, TRC
Produktkod. 30418649
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30418649 10 mg
1 options
Produktkod. 30418649 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68240510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,4-Secocholest-5-ene-3,4-dioic Acid Dimethyl Ester
CAS 55651-92-8
Molekylformel C29H48O4
Kvantitet 10 mg
Synonym 3-[(1R)-1,5-Dimethylhexyl]-2,3,3a,4,5,5a,6,9,9a,9b-decahydro-7-(methoxycarbonyl)-3a,6-dimethyl-1H-benz[e]indene-6-propanoic Acid Methyl Ester
InChI formel InChI=1S/C29H48O4/c1-19(2)9-8-10-20(3)22-13-14-23-21-11-12-25(27(31)33-7)29(5,18-16-26(30)32-6)24(21)15-17-28(22,23)4/h12,19-24H,8-11,13-18H2,1-7H3/t20-,21?,22?,23?,24?,28-,29-/m1/s1
LEDER COC(=O)CC[C@]1(C)C2CC[C@]3(C)C(CCC3C2CC=C1C(=O)OC)[C@H](C)CCCC(C)C
IUPAC-namn methyl (3aR,6R)-6-(3-methoxy-3-oxopropyl)-3a,6-dimethyl-3-[(2R)-6-methylheptan-2-yl]-2,3,4,5,5a,9,9a,9b-octahydro-1H-cyclopenta[a]naphthalene-7-carboxylate
Molekylvikt (g/mol) 460.69
Formel vikt 460.3553