Läs mer
3,5,3',5'-Tetraiodo Thyroacetamide, TRC
Produktkod. 30134583
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134583 100 mg
30011097 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134583 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT296120100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 176258-88-1
Molekylformel C14 H9 I4 N O3
Molekylvikt (g/mol) 746.84
Synonym Benzeneacetamide, 4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-
IUPAC-namn 2-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetamide
LEDER NC(=O)Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1
Specifikationer
Kemiskt namn eller material 2-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetamide
Kvantitet 100 mg
InChI formel InChI=1S/C14H9I4NO3/c15-8-4-7(5-9(16)13(8)21)22-14-10(17)1-6(2-11(14)18)3-12(19)20/h1-2,4-5,21H,3H2,(H2,19,20)
Formel vikt 746.6761