Läs mer
3,5,3',5'-Tetraiodo Thyroaldehyde, TRC
Produktkod. 30133807
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133807 100 mg
30133697 25 mg
30012423 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133807 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT296135100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2016-06-0
Molekylformel C13 H6 I4 O3
Molekylvikt (g/mol) 717.8
Synonym Benzaldehyde, 4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-, 4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzaldehyde, Imp I (EP)
IUPAC-namn 4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzaldehyde
LEDER Oc1c(I)cc(Oc2c(I)cc(C=O)cc2I)cc1I
Specifikationer
Kemiskt namn eller material 3,5,3',5'-Tetraiodo Thyroaldehyde
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C13H6I4O3/c14-8-3-7(4-9(15)12(8)19)20-13-10(16)1-6(5-18)2-11(13)17/h1-5,19H
Formel vikt 717.6496
Procent renhet >95%