Läs mer
3,5,6-Trichloro-2-pyridinol, TRC
Produktkod. 30134373
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134373 1 g
30135847 5 g
17951075 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134373 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7738601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6515-38-4
Molekylformel C5 H2 Cl3 N O
Molekylvikt (g/mol) 198.43
Synonym 3,5,6-Trichloro-2-pyridinol, 2(1H)-Pyridinone, 3,5,6-trichloro-, 2(1H)-Pyridone, 3,5,6-trichloro- (8CI), 2-Pyridinol, 3,5,6-trichloro- (7CI), 3,5,6-Trichloro-2(1H)-pyridinone, 2,3,5-Trichloro-6-hydroxypyridine, 2-Hydroxy-3,5,6-trichloropyridine, 2-Hydroxyl-3,5,6-trichloropyridine, 3,5,6-Trichloro-1H-pyridin-2-one, 3,5,6-Trichloro-2-hydroxypyridine, 3,5,6-Trichloropyridine-2-ol, TCP
IUPAC-namn 3,5,6-trichloropyridin-2-ol
LEDER Oc1nc(Cl)c(Cl)cc1Cl
Specifikationer
Kemiskt namn eller material 3,5,6-Trichloro-2-pyridinol
Kvantitet 1 g
InChI formel InChI=1S/C5H2Cl3NO/c6-2-1-3(7)5(10)9-4(2)8/h1H,(H,9,10)
Formel vikt 196.9202