missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(1,1-dimethylethyl)-1-hydroxy-4-oxo-2,5-cyclohexadiene-1-propanoic Acid, TRC

Product Code. 30011454
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
2.5 mg
10 mg
25 mg
missing translation for 'unitSize'
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30011454 2.5 mg 2.5mg
30004084 25 mg -
30006909 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30011454 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB4296502.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83237-15-4
Molekylformel C17 H26 O4
Molekylvikt (g/mol) 294.39
Synonym 2,5-Cyclohexadiene-1-propanoic acid, 3,5-bis(1,1-dimethylethyl)-1-hydroxy-4-oxo-, 3,5-Bis(1,1-dimethylethyl)-1-hydroxy-4-oxo-2,5-cyclohexadiene-1-propanoic acid, 3-(3,5-di-tert-Butyl-1-hydroxy-4-oxocyclohexa-2,5-dien-1-yl)propanoic acid
IUPAC-namn 3-(3,5-ditert-butyl-1-hydroxy-4-oxocyclohexa-2,5-dien-1-yl)propanoic acid
LEDER CC(C)(C)C1=CC(O)(CCC(=O)O)C=C(C1=O)C(C)(C)C

Specifications

Kemiskt namn eller material 3,5-Bis(1,1-dimethylethyl)-1-hydroxy-4-oxo-2,5-cyclohexadiene-1-propanoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C17H26O4/c1-15(2,3)11-9-17(21,8-7-13(18)19)10-12(14(11)20)16(4,5)6/h9-10,21H,7-8H2,1-6H3,(H,18,19)
Formel vikt 294.1831
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.