missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzamidoxime, 97%

Brand:  Alfa Aesar™ H64759.03

Additional Details : CAS Number : 72111-09-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15492089

  • 1947.58 SEK / 1g


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Bis(trifluoromethyl)benzamidoxime
72111-09-2
MFCD00068188

Specifications

3,5-Bis(trifluoromethyl)benzamidoxime
72111-09-2
97%
MFCD00068188
1g
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=NO)N
7330429
272.15
Solid
C9H6F6N2O
129° to 132°C (lit.)
GGAPMNUHESBFNK-UHFFFAOYSA-N
272.15
Videos
SDS