Läs mer
3-[[(5-Carboxypentyl)amino]carbonyl]-3-hydroxypentanedioic Acid, TRC
Produktkod. 30354222
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354222 100 mg
30363799 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30354222 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC181165100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-[((5-Carboxypentyl)amino)carbonyl]-3-hydroxypentanedioic Acid
CAS 1675217-43-2
Molekylformel C12H19NO8
Kvantitet 100 mg
InChI formel InChI=1S/C12H19NO8/c14-8(15)4-2-1-3-5-13-11(20)12(21,6-9(16)17)7-10(18)19/h21H,1-7H2,(H,13,20)(H,14,15)(H,16,17)(H,18,19)
LEDER OC(=O)CCCCCNC(=O)C(O)(CC(=O)O)CC(=O)O
IUPAC-namn 3-(5-carboxypentylcarbamoyl)-3-hydroxypentanedioic acid
Molekylvikt (g/mol) 305.28
Formel vikt 305.1111