Läs mer
3,5-Di-tert-butyl-4-hydroxybenzoic acid hexadecyl ester, TRC
Produktkod. 30373595
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30373595 100 mg
1 options
Produktkod. 30373595 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD493733100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,5-Di-tert-butyl-4-hydroxybenzoic acid hexadecyl ester
CAS 67845-93-6
Molekylformel C31H54O3
Kvantitet 100 mg
Synonym 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzoic acid hexadecyl ester, 3,5-Di-tert-butyl-4-hydroxybenzoic acid hexadecyl ester, Antioxidant 2908, Cyagard UV 2908, Cyasorb 2908, Cyasorb UV 2908, Hexadecyl 3,5-bis-tert-butyl-4-hydroxybenzoate, Hexadecyl 3,5-di-tert-butyl-4-hydroxybenzoate, Hexadecyl 4-hydroxy-3,5-di(tert-butyl)benzoate, Kemisorb 114, Light Stabilizer 2908, Palmityl 3,5-di-tert-butyl-4-hydroxybenzoate, Sabostab UV 37, Songsorb 2908PW, UV 2908, 3,5-Di-tert-butyl-4-hydroxybenzoic acid-hexadecyl ester, 3,5-Di-tert-butyl-4-hydroxybenzoic Acid-Hexadecyl Ester
InChI formel InChI=1S/C31H54O3/c1-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-34-29(33)25-23-26(30(2,3)4)28(32)27(24-25)31(5,6)7/h23-24,32H,8-22H2,1-7H3
LEDER CCCCCCCCCCCCCCCCOC(C1=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C1)=O
Molekylvikt (g/mol) 474.76