Läs mer
3,5-Diamino-6-chloropyrazine-2-carboxylic Acid Methyl Ester, TRC
Produktkod. 30367280
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30367280 2.5 g
1 options
Produktkod. 30367280 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4162402.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3,5-Diamino-6-chloropyrazine-2-carboxylate
CAS 1458-01-1
Molekylformel C6 H7 Cl N4 O2
Kvantitet 2.5 g
Synonym Methyl 3,5-Diamino-6-chloropyrazine-2-carboxylate, Amiloride Related Compound A (USP), 3,5-Diamino-6-chloropyrazine-2-carboxylic Acid Methyl Ester, Amiloride impurity A (EP)
InChI formel InChI=1S/C6H7ClN4O2/c1-13-6(12)2-4(8)11-5(9)3(7)10-2/h1H3,(H4,8,9,11)
LEDER COC(=O)c1nc(Cl)c(N)nc1N
IUPAC-namn methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
Molekylvikt (g/mol) 202.6
Formel vikt 202.0258