missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture, Thermo Scientific Chemicals
216.17 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
100 g
500 g
Description
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | ∼235°C (decomposition) |
Flash Point | 210°C (410°F) |
Quantity | 50 g |
Beilstein | 2086484 |
Merck Index | 14,2978 |
Solubility Information | Very soluble in water. |
Formula Weight | 152.15 |
Percent Purity | 98% |
Assay | may contain up to 3% moisture |
Chemical Name or Material | 3,5-Diaminobenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'