missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromoaniline, 98%, Thermo Scientific Chemicals

Catalog Number 15519533
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
626-40-4
C6H5Br2N
250.92
RVNUUWJGSOHMRR-UHFFFAOYSA-N
3,5-dibromophenylamine, 3,5-dibromanilin, benzenamine, 3,5-dibromo, 1-amino-3,5-dibromobenzene, 3,5-dibromobenzeneamine, 3,5-dibromoamiline, 3,5-di-bromo-aniline, 3,5-dibromo-phenylamine, acmc-1b0ij, ksc493e8l
221512
3,5-dibromoaniline
C1=C(C=C(C=C1Br)Br)N
This item is not returnable. View return policy
626-40-4
C6H5Br2N
250.92
RVNUUWJGSOHMRR-UHFFFAOYSA-N
3,5-dibromophenylamine, 3,5-dibromanilin, benzenamine, 3,5-dibromo, 1-amino-3,5-dibromobenzene, 3,5-dibromobenzeneamine, 3,5-dibromoamiline, 3,5-di-bromo-aniline, 3,5-dibromo-phenylamine, acmc-1b0ij, ksc493e8l
221512
3,5-dibromoaniline
C1=C(C=C(C=C1Br)Br)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 53.0°C to 54.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 250.92
Percent Purity 98%
Chemical Name or Material 3,5-Dibromoaniline
compliance-icons
Product Identifier
  • 3,5-Dibromoaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'