missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromosalicylaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-103040
Click to view available options
Quantity:
1 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
90-59-5
MFCD00003318
JHZOXYGFQMROFJ-UHFFFAOYSA-N
3,5-dibromosalicylaldehyde, dalyde, 3,5-dibromosalicylaldeyde, benzaldehyde, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzaldehyde, 3,5-dibromosilicylaldehyde, unii-7a260pbi3a, salicylaldehyde, 3,5-dibromo, 3,5-dibromsalicylaldehyd, 3,5-dibromo salicyclic aldehyde
7024
3,5-dibromo-2-hydroxybenzaldehyde
C1=C(C=C(C(=C1Br)O)C=O)Br
This item is not returnable. View return policy
90-59-5
MFCD00003318
JHZOXYGFQMROFJ-UHFFFAOYSA-N
3,5-dibromosalicylaldehyde, dalyde, 3,5-dibromosalicylaldeyde, benzaldehyde, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzaldehyde, 3,5-dibromosilicylaldehyde, unii-7a260pbi3a, salicylaldehyde, 3,5-dibromo, 3,5-dibromsalicylaldehyd, 3,5-dibromo salicyclic aldehyde
7024
3,5-dibromo-2-hydroxybenzaldehyde
C1=C(C=C(C(=C1Br)O)C=O)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 80°C to 84°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H4Br2O2
Linear Formula Br2C6H2(OH)CHO
Quantity 1 g
Beilstein 08,55
Merck Index 15,3032
Molecular Weight (g/mol) 279.91
Formula Weight 279.91
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dibromosalicylaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dibromosalicylaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'