Learn More
3,5-Dibromothiophene-2-carboxylic acid, 97%, ACROS Organics™
Brand:  Acros Organics 446470010
Additional Details : CAS Number : 7311-68-4 Weight : 0.05100kg
Quantity 1g
 View more versions of this product
Product Code. 12163751
  • 768.00 SEK / 1g
Estimated Shipment: 17-08-2021
Specifications
Chemical Identifiers
7311-68-4
285.94
YWOSIPRAZKDEIL-UHFFFAOYSA-N
625455
OC(=O)C1=C(Br)C=C(Br)S1
C5H2Br2O2S
MFCD01006744
3,5-dibromo-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 3,5-dibromo, 3,5-dibromo-thiophene-2-carboxylic acid, 3,5-dibromo-2-thiophenecarboxylic acid #, 3,5-dibromothiophene-2-carboxylic acid
3,5-dibromothiophene-2-carboxylic acid
Specifications
3, 5-Dibromothiophene-2-carboxylic acid
180°C to 188°C
7311-68-4
C5H2Br2O2S
3,5-dibromo-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 3,5-dibromo, 3,5-dibromo-thiophene-2-carboxylic acid, 3,5-dibromo-2-thiophenecarboxylic acid #, 3,5-dibromothiophene-2-carboxylic acid
OC(=O)C1=C(Br)C=C(Br)S1
285.94
285.94
Glass Bottle
1g
97%
MFCD01006744
YWOSIPRAZKDEIL-UHFFFAOYSA-N
3,5-dibromothiophene-2-carboxylic acid
625455
97%
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 3,5-Dibromothiophene-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.