Läs mer
3,5-Dichloro-2,6-dimethylpyridin-4-amine, Mikromol
Produktkod. 30458278
Change view
Kvantitet:
100 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30458278 4 x 25 mg
17921276 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30458278 Leverantör Mikromol Leverantörsnummer MM0699.0100254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3,5-Dichloro-2,6-dimethylpyridin-4-amine, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,5-Dichloro-2,6-dimethylpyridin-4-amine
CAS 50978-40-0
Analyt- eller komponentnamn 3,5-Dichloro-2,6-dimethylpyridin-4-amine
Frakt skick Room Temperature
Molekylformel C7 H8 Cl2 N2
Certifikat och efterlevnad ISO 9001
Synonym 3,5-Dichloro-2,6-dimethyl-4-pyridinamine, 4-Amino-3,5-dichloro-2,6-lutidine
InChI formel InChI=1S/C7H8Cl2N2/c1-3-5(8)7(10)6(9)4(2)11-3/h1-2H3,(H2,10,11)
Lösningstyp Neat
LEDER Cc1nc(C)c(Cl)c(N)c1Cl
IUPAC-namn 3,5-dichloro-2,6-dimethylpyridin-4-amine
Molekylvikt (g/mol) 191.06
Kvantitet 4 x 25 mg
Formel vikt 190.0065 g/mol