Läs mer
3,5-Dichloro-2-methyl-benzoquinone, TRC
Produktkod. 17974705
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17974705 50 mg
17971385 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17974705 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43535550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 40100-98-9
Molekylformel C7 H4 Cl2 O2
Molekylvikt (g/mol) 191.011
Synonym 3,5-Dichloro-2-methyl-2,5-cyclohexadiene-1,4-dione
IUPAC-namn 3,5-dichloro-2-methyl-1,4-benzoquinone
LEDER CC1=C(Cl)C(=O)C(=CC1=O)Cl
Specifikationer
Kemiskt namn eller material 3,5-Dichloro-2-methyl-benzoquinone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C7H4Cl2O2/c1-3-5(10)2-4(8)7(11)6(3)9/h2H,1H3
Formel vikt 189.9588
Procent renhet >95%