Learn More
3,5-Dichloro-4-hydroxybenzaldehyde, 97%, Thermo Scientific™
Catalog Number p-3485394
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2314-36-5
C7H4Cl2O2
191.007
MFCD00017605
LIYGCLJYTHRBQV-UHFFFAOYSA-N
benzaldehyde, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzaldehyde, benzaldehyde,3,5-dichloro-4-hydroxy, 2,6-dichloro-4-formylphenol, benzaldehyde,5-dichloro-4-hydroxy, 1-formyl-3,5-dichloro-4-hydroxy-benzene, 3,5-bis chloranyl-4-oxidanyl-benzaldehyde
16839
3,5-dichloro-4-hydroxybenzaldehyde
C1=C(C=C(C(=C1Cl)O)Cl)C=O
2314-36-5
C7H4Cl2O2
191.007
MFCD00017605
LIYGCLJYTHRBQV-UHFFFAOYSA-N
benzaldehyde, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzaldehyde, benzaldehyde,3,5-dichloro-4-hydroxy, 2,6-dichloro-4-formylphenol, benzaldehyde,5-dichloro-4-hydroxy, 1-formyl-3,5-dichloro-4-hydroxy-benzene, 3,5-bis chloranyl-4-oxidanyl-benzaldehyde
16839
3,5-dichloro-4-hydroxybenzaldehyde
C1=C(C=C(C(=C1Cl)O)Cl)C=O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 191.01
Percent Purity 97%
Chemical Name or Material 3, 5-Dichloro-4-hydroxybenzaldehyde