missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzenethiol, 97%, Thermo Scientific Chemicals

Catalog Number p-4258937
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
17231-94-6
C6H4Cl2S
179.07
WRXIPCQPHZMXOO-UHFFFAOYSA-N
3,5-dichlorothiophenol, 3,5-dichloro thiophenol, 3,5-dichlorobenzenthiol, 3,5-dichlorobenzene-1-thiol, pubchem6804, 3,5-dichloro-thiophenol, 3,5-dichloro-benzenethiol, acmc-209e4e, 3,5-dichlorophenyl mercaptan, benzenethiol, 3,5-dichloro
2736096
3,5-dichlorobenzenethiol
C1=C(C=C(C=C1Cl)Cl)S
This item is not returnable. View return policy
17231-94-6
C6H4Cl2S
179.07
WRXIPCQPHZMXOO-UHFFFAOYSA-N
3,5-dichlorothiophenol, 3,5-dichloro thiophenol, 3,5-dichlorobenzenthiol, 3,5-dichlorobenzene-1-thiol, pubchem6804, 3,5-dichloro-thiophenol, 3,5-dichloro-benzenethiol, acmc-209e4e, 3,5-dichlorophenyl mercaptan, benzenethiol, 3,5-dichloro
2736096
3,5-dichlorobenzenethiol
C1=C(C=C(C=C1Cl)Cl)S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 62°C to 65°C
Color White to Yellow
Boiling Point 270°C to 272°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3SH
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 179.07
Percent Purity 97%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 3, 5-Dichlorobenzenethiol
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dichlorobenzenethiol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'