missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-2-methoxybenzyl bromide, 97%

Brand:  Alfa Aesar™ H26582.03

Additional Details : CAS Number : 886500-63-6 Weight : 0.00100kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11446040

  • 1222.81 SEK / 1g


Estimated Shipment
24-08-2020
Add to basket

Specifications

Chemical Identifiers

886500-63-6
237.044
XLYRQPGSEJMVIM-UHFFFAOYSA-N
17750690
COC1=C(C=C(C=C1CBr)F)F
C8H7BrF2O
MFCD06660240
1-bromomethyl-3,5-difluoro-2-methoxybenzene, 3,5-difluoro-2-methoxybenzyl bromide, 3,5-difluoro-2-methoxybenzylbromide, benzene,1-bromomethyl-3,5-difluoro-2-methoxy, 1-bromomethyl-3,5-bis fluoranyl-2-methoxy-benzene
1-(bromomethyl)-3,5-difluoro-2-methoxybenzene

Specifications

3,5-Difluoro-2-methoxybenzyl bromide
886500-63-6
MFCD06660240
1-bromomethyl-3,5-difluoro-2-methoxybenzene, 3,5-difluoro-2-methoxybenzyl bromide, 3,5-difluoro-2-methoxybenzylbromide, benzene,1-bromomethyl-3,5-difluoro-2-methoxy, 1-bromomethyl-3,5-bis fluoranyl-2-methoxy-benzene
COC1=C(C=C(C=C1CBr)F)F
237.044
237.04
1g
C8H7BrF2O
UN3265
XLYRQPGSEJMVIM-UHFFFAOYSA-N
1-(bromomethyl)-3,5-difluoro-2-methoxybenzene
17750690
97%
Videos
SDS Documents