missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluoro-4-methoxybenzyl bromide, 97%, Thermo Scientific Chemicals
812.62 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
Specifications
Melting Point | 39°C to 42°C |
Quantity | 1 g |
UN Number | UN3261 |
Solubility Information | Insoluble in water. |
Formula Weight | 237.04 |
Percent Purity | 97% |
Chemical Name or Material | 3,5-Difluoro-4-methoxybenzyl bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'