missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(3-fluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62456.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15497456

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

MFCD17629840

Specifications

3,5-Difluoro-N-(3-fluorophenyl)benzenesulfonamide
MFCD17629840
Solid
5g
287.26
97%
Videos
SDS Documents