missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(3-methylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62597.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477616

  • 2834.60 SEK / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-(3-methylphenyl)benzenesulfonamide
283.29
N-(m-Tolyl)-3,5-difluorobenzenesulfonamide
C13H11F2NO2S
MFCD17629715

Specifications

3,5-Difluoro-N-(3-methylphenyl)benzenesulfonamide
97%
283.29
N-(m-Tolyl)-3,5-difluorobenzenesulfonamide
1g
C13H11F2NO2S
MFCD17629715
Videos
SDS