Läs mer
3,5-Difluorobenzoic acid, 97%, Thermo Scientific™
Produktkod. 11468886
Change view
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11468886 5 g 5g
11478886 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11468886 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A17033.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 455-40-3
Molekylformel C7H4F2O2
Molekylvikt (g/mol) 158.104
MDL-nummer MFCD00010323
InChI-nyckel GONAVIHGXFBTOZ-UHFFFAOYSA-N
Synonym 3,5-difluorobenzoicacid, benzoic acid, 3,5-difluoro, pubchem2173, 3,5 difluorobenzoic acid, 3,5-difluoro benzoicacid, acmc-209k3a, 3,5 difluoro benzoic acid, 3,5-di-fluorobenzoic acid, 3,5-difluoro-benzoic acid
PubChem CID 101265
IUPAC-namn 3,5-difluorobenzoic acid
LEDER C1=C(C=C(C=C1F)F)C(=O)O
Specifikationer
Kemiskt namn eller material 3,5-Difluorobenzoic acid
Smältpunkt 119°C to 123°C
Odör Odorless
Kvantitet 5 g
Beilstein 1940680
Formel vikt 158.11
Procent renhet 97%