Läs mer
3,5-Difluorobenzoic Acid, TRC
Produktkod. 30133233
Change view
Kvantitet:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133233 25 g
30012174 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133233 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45115825G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 455-40-3
Molekylformel C7H4F2O2
Molekylvikt (g/mol) 158.1
Synonym NSC 403726, 3,5-Difluoro-benzoic Acid
LEDER FC1=CC(F)=CC(C(O)=O)=C1
Specifikationer
Kemiskt namn eller material 3,5-Difluorobenzoic Acid
Kvantitet 25 g
InChI formel InChI=1S/C7H4F2O2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3H,(H,10,11)
Formel vikt 158.0179