missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoropyridine, 97%, ACROS Organics™

Brand:  Acros Organics 442690010

Additional Details : CAS Number : 71902-33-5 Weight : 0.01000kg Transport : UN number : 1993 Chem class : 3 Pack group : II


Quantity 1g

 View more versions of this product

Product Code. 15204238

  • 393.66 SEK / 1g


Estimated Shipment
03-11-2020
Add to basket


Specifications

Chemical Identifiers

71902-33-5
115.08
pyridine, 3,5-difluoro, 3,5-difluoro-pyridine, abbypharma ap-14-5380, 3,5-difluoropyridine;, pubchem10556, acmc-209okl, 3,5-difluoro-pyridine;, ksc377a3n, 3,5-difluoropyridine, 3,5-difluoro pyridine
3,5-difluoropyridine
C5H3F2N
WRXAZPPGFLETFR-UHFFFAOYSA-N
642820
C1=C(C=NC=C1F)F

Specifications

3,5-Difluoropyridine
7°C
Glass Bottle
92.0°C to 94.0°C
71902-33-5
C5H3F2N
WRXAZPPGFLETFR-UHFFFAOYSA-N
3,5-difluoropyridine
642820
97%
1.2560g/mL
Authentic
1.256
1g
96% min. (GC)
pyridine, 3,5-difluoro, 3,5-difluoro-pyridine, abbypharma ap-14-5380, 3,5-difluoropyridine;, pubchem10556, acmc-209okl, 3,5-difluoro-pyridine;, ksc377a3n, 3,5-difluoropyridine, 3,5-difluoro pyridine
C1=C(C=NC=C1F)F
115.08
115.08
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,5-Difluoropyridine, 97%

Signal Word
  • Danger

Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents