missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoropyridine, 97%, Thermo Scientific Chemicals

Catalog Number 15214238
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
71902-33-5
C5H3F2N
115.08
WRXAZPPGFLETFR-UHFFFAOYSA-N
pyridine, 3,5-difluoro, 3,5-difluoro-pyridine, abbypharma ap-14-5380, 3,5-difluoropyridine;, pubchem10556, acmc-209okl, 3,5-difluoro-pyridine;, ksc377a3n, 3,5-difluoropyridine, 3,5-difluoro pyridine
642820
3,5-difluoropyridine
C1=C(C=NC=C1F)F
This item is not returnable. View return policy
71902-33-5
C5H3F2N
115.08
WRXAZPPGFLETFR-UHFFFAOYSA-N
pyridine, 3,5-difluoro, 3,5-difluoro-pyridine, abbypharma ap-14-5380, 3,5-difluoropyridine;, pubchem10556, acmc-209okl, 3,5-difluoro-pyridine;, ksc377a3n, 3,5-difluoropyridine, 3,5-difluoro pyridine
642820
3,5-difluoropyridine
C1=C(C=NC=C1F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.2560g/mL
Boiling Point 92.0°C to 94.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.256
Formula Weight 115.08
Percent Purity 97%
Chemical Name or Material 3,5-Difluoropyridine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,5-Difluoropyridine, 97%
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'