Läs mer
3,5-Difluorothiophenol, TRC
Produktkod. 30378946
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378946 100 mg
30370775 500 mg
30387087 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30378946 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD452080100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99389-26-1
Molekylformel C6H4F2S
Molekylvikt (g/mol) 146.15
IUPAC-namn 3,5-difluorobenzenethiol
LEDER SC1=CC(F)=CC(F)=C1
Specifikationer
Kemiskt namn eller material 3,5-Difluorothiophenol
Kvantitet 100 mg
InChI formel InChI=1S/C6H4F2S/c7-4-1-5(8)3-6(9)2-4/h1-3,9H
Formel vikt 146