Läs mer
3,5-Dihydroxy-4-methoxybenzoic Acid, TRC
Produktkod. 30136089
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136089 250 mg
30007812 50 mg
30007191 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136089 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD496360250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4319-02-2
Molekylformel C8 H8 O5
Molekylvikt (g/mol) 184.15
Synonym Benzoic acid, 3,5-dihydroxy-4-methoxy-, α-Resorcylic acid, 4-methoxy- (6CI,7CI,8CI), 3,5-Dihydroxy-4-methoxybenzoic acid, 4-Methoxy-3,5-dihydroxybenzoic acid, 4-O-Methylgallic acid, Gallic acid 4-methyl ether, 5-Hydroxyisovanillic acid
IUPAC-namn 3,5-dihydroxy-4-methoxybenzoic acid
LEDER COc1c(O)cc(cc1O)C(=O)O
Specifikationer
Kemiskt namn eller material 5-Hydroxyisovanillic Acid
Kvantitet 250 mg
InChI formel InChI=1S/C8H8O5/c1-13-7-5(9)2-4(8(11)12)3-6(7)10/h2-3,9-10H,1H3,(H,11,12)
Formel vikt 184.0372