Läs mer
3,5-Diiodo-4-hydroxyphenylacetic Acid Ethyl Ester, TRC
Produktkod. 30375132
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375132 250 mg
30371606 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30375132 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD455270250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90917-49-0
Molekylformel C10H10I2O3
Molekylvikt (g/mol) 431.99
Synonym (4-Hydroxy-3,5-diiodophenyl)acetic Acid Ethyl Ester, 4-Hydroxy-3,5-diiodo-benzeneacetic Acid Ethyl Ester
LEDER CCOC(=O)Cc1cc(I)c(O)c(I)c1
Specifikationer
Kemiskt namn eller material 3,5-Diiodo-4-hydroxyphenylacetic Acid Ethyl Ester
Kvantitet 250 mg
InChI formel InChI=1S/C10H10I2O3/c1-2-15-9(13)5-6-3-7(11)10(14)8(12)4-6/h3-4,14H,2,5H2,1H3
Formel vikt 431.8719