Läs mer
3’,5’-Diiodo-L-thyronine, TRC
Produktkod. 30135876
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135876 250 mg
30007576 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135876 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD455156250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4192-14-7
Molekylformel C15H13I2NO4
Molekylvikt (g/mol) 525.08
Synonym (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)phenyl)propanoic Acid, 3',5'-Diiodothyronine, O-(4-Hydroxy-3,5-diiodophenyl)-L-tyrosine, 3-[p-(4-Hydroxy-3,5-diiodophenoxy)phenyl]-L-alanine
LEDER OC(C(I)=C1)=C(I)C=C1OC(C=C2)=CC=C2C[C@H](N)C(O)=O
Specifikationer
Kemiskt namn eller material 3',5'-Diiodo-L-thyronine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C15H13I2NO4/c16-11-6-10(7-12(17)14(11)19)22-9-3-1-8(2-4-9)5-13(18)15(20)21/h1-4,6-7,13,19H,5,18H2,(H,20,21)/t13-/m0/s1
Kvantitet 250 mg
Formel vikt 524.8934
Procent renhet >95%