Learn More
3,5-Diiodosalicylaldehyde, 97%, Acros Organics
435.49 SEK - 1416.96 SEK
Chemical Identifiers
CAS 2631-77-8
Molecular Formula C7H4I2O2
Molecular Weight (g/mol) 373.92
InChI Key MYWSBJKVOUZCIA-UHFFFAOYSA-N
Synonym 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo
PubChem CID 75829
IUPAC Name 2-hydroxy-3,5-diiodobenzaldehyde
SMILES C1=C(C=C(C(=C1I)O)C=O)I
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15209676 View Documents Acros Organics
450860050
5g 435.49 SEK
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 11-06-2020
Add to basket
15219676 View Documents Acros Organics
450860250
25g 1416.96 SEK
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 11-06-2020
Add to basket
Specifications
Chemical Identifiers
2631-77-8
373.92
3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo
2-hydroxy-3,5-diiodobenzaldehyde
C7H4I2O2
MYWSBJKVOUZCIA-UHFFFAOYSA-N
75829
C1=C(C=C(C(=C1I)O)C=O)I
Specifications
3, 5-Diiodosalicylaldehyde
5g
96% min. (GC)
3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo
C1=C(C=C(C(=C1I)O)C=O)I
373.92
373.92
108°C to 110°C
2631-77-8
C7H4I2O2
MYWSBJKVOUZCIA-UHFFFAOYSA-N
2-hydroxy-3,5-diiodobenzaldehyde
75829
97%
Safety and Handling
Safety and Handling
Product Identifier
  • 3,5-Diiodosalicylaldehyde, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.